accessibility menu, dialog, popup

UserName

CAS RN 1744-43-0

IUPAC Name: 2-fluoro-5-(trifluoromethyl)benzene-1-sulfonyl chloride
Synonyms: 2-fluoro-5-(trifluoromethyl)benzenesulfonyl chloride
Molecular Weight (g/mol): 262.6
Molecular Formula: C7H3ClF4O2S
InChi Key: XTHDSFNKYHMRFZ-UHFFFAOYSA-N
SMILES: FC1=C(C=C(C=C1)C(F)(F)F)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results