missing translation for 'onlineSavingsMsg'
Learn More

2'-Fluoro-2'-deoxyadenosine, 99%, Thermo Scientific Chemicals

Product Code. p-7043081
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 15486899

Brand: Thermo Scientific Alfa Aesar J65441.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2'-Fluoro-2'-deoxyadenosine is a potential prodrug for 2-fluroadenine, involves deoxygenation by reduction of the 2'-thiocar-bonylimidazolide with (Tms)3SiH as hydrogen donor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Fluoro-2′-deoxyadenosine is a potential prodrug for 2-fluroadenine, involves deoxygenation by reduction of the 2′-thiocar-bonylimidazolide with (Tms)3SiH as hydrogen donor.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64183-27-3
Molecular Formula C10H12FN5O3
Molecular Weight (g/mol) 269.24
MDL Number MFCD09750859
InChI Key ZGYYPTJWJBEXBC-GPGUAWMRNA-N
Synonym 2'-fluoro-2'-deoxyadenosine, 2'-deoxy-2'-fluoroadenosine, adenosine, 2'-deoxy-2'-fluoro, 2r,3r,4r,5r-5-6-amino-9h-purin-9-yl-4-fluoro-2-hydroxymethyl tetrahydrofuran-3-ol, 9-2-fluoro-2-deoxy-beta-d-ribofuranosyl-purin-6-amine, 2r,3r,4r,5r-5-6-aminopurin-9-yl-4-fluoro-2-hydroxymethyl oxolan-3-ol, 2r,3r,4r,5r-5-6-aminopurin-9-yl-4-fluoro-2-hydroxymethyl tetrahydrofuran-3-ol, 2-fluoro-2-deoxyadenosine, 2'f-da
PubChem CID 100253
SMILES NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3F)C2=NC=N1

Specifications

Color White
Quantity 1 g
Solubility Information Soluble in water.
IUPAC Name (2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-4-fluoro-2-(hydroxymethyl)oxolan-3-ol
Formula Weight 269.23
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2'-Fluoro-2'-deoxyadenosine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.