missing translation for 'onlineSavingsMsg'
Learn More

2-(Ethylthio)nicotinic acid, 98%, Thermo Scientific™

Product Code. p-7044690
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11332057

Brand: Thermo Scientific Alfa Aesar L07801.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27868-76-4
Molecular Formula C8H9NO2S
Molecular Weight (g/mol) 183.225
MDL Number MFCD00051680
InChI Key LYHPPDODTUEXAN-UHFFFAOYSA-N
Synonym 2-ethylthio nicotinic acid, 2-ethylsulfanyl pyridine-3-carboxylic acid, 2-ethylthio pyridine-3-carboxylic acid, 2-ethylthiopyridine-3-carboxylic acid, maybridge1_001449, acmc-1cr2k, 2-ethylsulfanyl-nicotinic acid, 2-ethylsulfanyl nicotinic acid #, 3-pyridinecarboxylicacid, 2-ethylthio
PubChem CID 591734
IUPAC Name 2-ethylsulfanylpyridine-3-carboxylic acid
SMILES CCSC1=C(C=CC=N1)C(=O)O

Specifications

Melting Point 183°C to 187°C
Quantity 1 g
Beilstein 388961
Formula Weight 183.23
Percent Purity 98%
Chemical Name or Material 2-(Ethylthio)nicotinic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.