accessibility menu, dialog, popup

UserName

CAS RN 27868-76-4

CH3SNOOH

CAS RN 27868-76-4

IUPAC Name: 2-(ethylsulfanyl)pyridine-3-carboxylic acid
Synonyms: 2-(ethylsulfanyl)pyridine-3-carboxylic acid
Molecular Weight (g/mol): 183.23
Molecular Formula: C8H9NO2S
InChi Key: LYHPPDODTUEXAN-UHFFFAOYSA-N
SMILES: CCSC1=C(C=CC=N1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results