missing translation for 'onlineSavingsMsg'
Learn More

2-(Chloromethyl)phenyl isocyanate, 97%, Thermo Scientific Chemicals

Product Code. 11352837
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11352837 1 g 1g
11362837 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352837 Supplier Thermo Scientific Alfa Aesar Supplier No. L15995.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 52986-66-0 | Molecular Formula: C8H6ClNO | Molecular Weight: 167.592 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52986-66-0
Molecular Formula C8H6ClNO
Molecular Weight (g/mol) 167.592
MDL Number MFCD00077427
InChI Key XTYOTPBIDRGAML-UHFFFAOYSA-N
Synonym 2-chloromethyl phenyl isocyanate, 1-chloromethyl-2-isocyanatobenzene, acmc-20aono, 2-chloromethyl phenylisocyanate
PubChem CID 3462650
IUPAC Name 1-(chloromethyl)-2-isocyanatobenzene
SMILES C1=CC=C(C(=C1)CCl)N=C=O

Specifications

Boiling Point 43°C to 45°C (0.2 mmHg)
Flash Point >110°C (230°F)
Quantity 1 g
UN Number UN2922
Beilstein 957161
Formula Weight 167.6
Percent Purity 97%
Chemical Name or Material 2-(Chloromethyl)phenyl isocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.