accessibility menu, dialog, popup

UserName

ClNCO

CAS RN 52986-66-0

IUPAC Name: 1-(chloromethyl)-2-isocyanatobenzene
Synonyms: 1-(chloromethyl)-2-isocyanatobenzene
Molecular Weight (g/mol): 167.59
Molecular Formula: C8H6ClNO
InChi Key: XTYOTPBIDRGAML-UHFFFAOYSA-N
SMILES: ClCC1=CC=CC=C1N=C=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results