missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-fluoro-3-methylbenzoyl chloride, 97%, Thermo Scientific™

Product Code. 11456946
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11456946 5 g 5g
11446946 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11456946 Supplier Thermo Scientific Alfa Aesar Supplier No. B24028.06
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 261762-82-7
Molecular Formula C8H5Cl2FO
Molecular Weight (g/mol) 207.025
MDL Number MFCD01631361
InChI Key XJFKTIHKOKWAEF-UHFFFAOYSA-N
Synonym 2-chloro-6-fluoro-3-methylbenzoylchloride, acmc-20anyo, benzoyl chloride,6-chloro-2-fluoro-3-methyl, 2-chloranyl-6-fluoranyl-3-methyl-benzoyl chloride, benzoyl chloride, 2-chloro-6-fluoro-3-methyl-9ci
PubChem CID 2773670
IUPAC Name 2-chloro-6-fluoro-3-methylbenzoyl chloride
SMILES CC1=C(C(=C(C=C1)F)C(=O)Cl)Cl

Specifications

Refractive Index 1.526
Quantity 5 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 207.03
Percent Purity 97%
Chemical Name or Material 2-Chloro-6-fluoro-3-methylbenzoyl chloride
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.