accessibility menu, dialog, popup

UserName

CH3FClOCl

CAS RN 261762-82-7

IUPAC Name: 2-chloro-6-fluoro-3-methylbenzoyl chloride
Synonyms: 2-chloro-6-fluoro-3-methylbenzoyl chloride
Molecular Weight (g/mol): 207.03
Molecular Formula: C8H5Cl2FO
InChi Key: XJFKTIHKOKWAEF-UHFFFAOYSA-N
SMILES: CC1=CC=C(F)C(C(Cl)=O)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results