missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-fluoro-3-methylbenzoyl chloride, 97%, Thermo Scientific™

Product Code. 11446946
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11446946

Brand: Thermo Scientific Alfa Aesar B24028.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 261762-82-7
Molecular Formula C8H5Cl2FO
Molecular Weight (g/mol) 207.025
MDL Number MFCD01631361
InChI Key XJFKTIHKOKWAEF-UHFFFAOYSA-N
Synonym 2-chloro-6-fluoro-3-methylbenzoylchloride, acmc-20anyo, benzoyl chloride,6-chloro-2-fluoro-3-methyl, 2-chloranyl-6-fluoranyl-3-methyl-benzoyl chloride, benzoyl chloride, 2-chloro-6-fluoro-3-methyl-9ci
PubChem CID 2773670
IUPAC Name 2-chloro-6-fluoro-3-methylbenzoyl chloride
SMILES CC1=C(C(=C(C=C1)F)C(=O)Cl)Cl

Specifications

Refractive Index 1.526
Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 207.03
Percent Purity 97%
Chemical Name or Material 2-Chloro-6-fluoro-3-methylbenzoyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.