Learn More
2-Chloro-4-methoxypyridine, 99%, Thermo Scientific™
Product Code. 11328646
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 11328646
Brand: Thermo Scientific Alfa Aesar L17697.03
Request Bulk or Custom Format
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is an active pharmaceutical ingredient.
Solubility
Soluble in alcohol.
Notes
Store at room temperature. Keep away from strong oxidizing agents. Ensure proper ventilation.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 17228-69-2
Molecular Formula C6H6ClNO
Molecular Weight (g/mol) 143.57
MDL Number MFCD02093951
InChI Key PMTPFBWHUOWTNN-UHFFFAOYSA-N
Synonym 2-chloro-4-methoxy-pyridine, pyridine, 2-chloro-4-methoxy, zlchem 749, pubchem6204, 4-methoxy-2-chloropyridine, 2-chloro-4-methoxylpyridine, 2-chloro-4-methoxy pyridine, ksc494i4l, 2-chloro-4-methoxypyridine
PubChem CID 1991128
IUPAC Name 2-chloro-4-methoxypyridine
SMILES COC1=CC(=NC=C1)Cl
Specifications
Density 1.258
Boiling Point 110°C to 115°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.541
Quantity 1 g
Beilstein 1423379
Solubility Information Soluble in alcohol.
Formula Weight 143.57
Percent Purity 99%
Chemical Name or Material 2-Chloro-4-methoxypyridine