accessibility menu, dialog, popup

UserName

CAS RN 17228-69-2

H3CONCl

CAS RN 17228-69-2

IUPAC Name: 2-chloro-4-methoxypyridine
Synonyms: 2-chloro-4-methoxypyridine
Molecular Weight (g/mol): 143.57
Molecular Formula: C6H6ClNO
InChi Key: PMTPFBWHUOWTNN-UHFFFAOYSA-N
SMILES: COC1=CC(Cl)=NC=C1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)