missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-methoxyphenylacetonitrile, 95%

Product Code. 11371939
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11371939 5 g 5g
11361939 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11371939 Supplier Thermo Scientific Alfa Aesar Supplier No. H26021.06
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 170737-93-6 | C9H8ClNO | 181.62 g/mol

2-Chloro-4-methoxyphenylacetonitrile is used as a raw material and an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-4-methoxyphenylacetonitrile is used as a raw material and an intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 170737-93-6
Molecular Formula C9H8ClNO
Molecular Weight (g/mol) 181.62
MDL Number MFCD08445710
InChI Key UKYXNWLFQOJYNQ-UHFFFAOYSA-N
Synonym 2-2-chloro-4-methoxyphenyl acetonitrile, 2-chloro-4-methoxyphenyl acetonitrile, 2-chloro-4-methoxyphenylacetonitrile, acmc-20appn, pubchem11235, 2-chloro-4-methoxyphenylacetnitrile, benzeneacetonitrile, 2-chloro-4-methoxy, 2-2-chloranyl-4-methoxy-phenyl ethanenitrile
PubChem CID 22327506
IUPAC Name 2-(2-chloro-4-methoxyphenyl)acetonitrile
SMILES COC1=CC(Cl)=C(CC#N)C=C1

Specifications

Melting Point 41°C to 43°C
Quantity 5 g
UN Number UN3439
Solubility Information Insoluble in water.
Formula Weight 181.62
Percent Purity 95%
Chemical Name or Material 2-Chloro-4-methoxyphenylacetonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.