accessibility menu, dialog, popup

UserName

H3CONCl

CAS RN 170737-93-6

IUPAC Name: 2-(2-chloro-4-methoxyphenyl)acetonitrile
Synonyms: 2-(2-chloro-4-methoxyphenyl)acetonitrile
Molecular Weight (g/mol): 181.62
Molecular Formula: C9H8ClNO
InChi Key: UKYXNWLFQOJYNQ-UHFFFAOYSA-N
SMILES: COC1=CC(Cl)=C(CC#N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results