Learn More
2-Chloro-4-fluorobenzyl chloride, 96%, Thermo Scientific™
Product Code. 11343287
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. 11343287
Brand: Thermo Scientific Alfa Aesar L13850.22
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2-Chloro-4-fluorobenzyl chloride is used as a pharmaceutical intermediate.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 93286-22-7
Molecular Formula C7H5Cl2F
Molecular Weight (g/mol) 179.015
MDL Number MFCD00236026
InChI Key HMIAXVWVTBIGON-UHFFFAOYSA-N
Synonym 2-chloro-4-fluorobenzyl chloride, 2-chloro-1-chloromethyl-4-fluorobenzene, 2-chloro-4-fluorochlorobenzyl, 2-chloro-1-chloromethyl-4-fluoro-benzene, 2-chloro-4-fluorobenzylchloride, benzene, 2-chloro-1-chloromethyl-4-fluoro, alpha,2-dichloro-4-fluorotoluene, acmc-209rkz, ksc493q6n, paragos 500065
PubChem CID 2725064
IUPAC Name 2-chloro-1-(chloromethyl)-4-fluorobenzene
SMILES C1=CC(=C(C=C1F)Cl)CCl
Specifications
Refractive Index 1.534
Quantity 100 g
UN Number UN3265
Beilstein 9043128
Formula Weight 179.02
Percent Purity 96%
Chemical Name or Material 2-Chloro-4-fluorobenzyl chloride