accessibility menu, dialog, popup

UserName

CAS RN 93286-22-7

FClCl

CAS RN 93286-22-7

IUPAC Name: 2-chloro-1-(chloromethyl)-4-fluorobenzene
Synonyms: 2-chloro-1-(chloromethyl)-4-fluorobenzene
Molecular Weight (g/mol): 179.02
Molecular Formula: C7H5Cl2F
InChi Key: HMIAXVWVTBIGON-UHFFFAOYSA-N
SMILES: FC1=CC=C(CCl)C(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results