missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4,5-difluorobenzoic acid, 98%, Thermo Scientific™

Product Code. 11441207
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. Quantity unitSize
11441207 100 g 100g
11421207 5 g 5g
11431207 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 11441207

Brand: Thermo Scientific Alfa Aesar B24174.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110877-64-0
Molecular Formula C7H2ClF2O2
Molecular Weight (g/mol) 191.54
MDL Number MFCD00077479
InChI Key CGFMLBSNHNWJAW-UHFFFAOYSA-M
Synonym 2-chloro-4,5-difluoro-benzoic acid, 2'-chloro-4',5'-difluorobenzoic acid, benzoic acid, 2-chloro-4,5-difluoro, pubchem1339, ksc177o4l, rarechem al bo 1000, timtec-bb sbb003616, 2-chloro-4,5-difluorobenzoic acid, benzoicacid, 2-chloro-4,5-difluoro
PubChem CID 737171
SMILES [O-]C(=O)C1=CC(F)=C(F)C=C1Cl

Specifications

Melting Point 104°C to 107°C
Boiling Point 257°C to 259°C
Quantity 100 g
Beilstein 4247522
IUPAC Name 2-chloro-4,5-difluorobenzoate
Formula Weight 192.55
Percent Purity 98%
Chemical Name or Material 2-Chloro-4,5-difluorobenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.