CAS RN 110877-64-0
CAS RN 110877-64-0
IUPAC Name:
2-chloro-4,5-difluorobenzoate
Synonyms:
2-chloro-4,5-difluorobenzoate
Molecular Weight (g/mol):
191.54
Molecular Formula:
C7H2ClF2O2
InChi Key:
CGFMLBSNHNWJAW-UHFFFAOYSA-M
SMILES:
[O-]C(=O)C1=CC(F)=C(F)C=C1Cl
1
–
2
of
2
results
1
2-Chloro-4,5-difluorobenzoic acid, 98%, Thermo Scientific™
CAS: 110877-64-0 Molecular Formula: C7H2ClF2O2 Molecular Weight (g/mol): 191.54 MDL Number: MFCD00077479 InChI Key: CGFMLBSNHNWJAW-UHFFFAOYSA-M Synonym: 2-chloro-4,5-difluoro-benzoic acid,2'-chloro-4',5'-difluorobenzoic acid,benzoic acid, 2-chloro-4,5-difluoro,pubchem1339,ksc177o4l,rarechem al bo 1000,timtec-bb sbb003616,2-chloro-4,5-difluorobenzoic acid,benzoicacid, 2-chloro-4,5-difluoro PubChem CID: 737171 SMILES: [O-]C(=O)C1=CC(F)=C(F)C=C1Cl