Learn More
2'-Amino-2'-deoxyuridine, 98%, Thermo Scientific™
Product Code. 15497129
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15497129 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15497129 Supplier Thermo Scientific Alfa Aesar Supplier No. J65923.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2′-Amino-2′-deoxyuridine is used in the synthesis of anticode oligonucleotide, ribozyme and in antisense. It also acts as an intermediate and used in chemical research.
Solubility
Partly soluble in water.
Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 26889-39-4
Molecular Formula C9H13N3O5
Molecular Weight (g/mol) 243.219
MDL Number MFCD01317284
InChI Key LLIPTMWIZVIUSX-XVFCMESISA-N
Synonym 2'-amino-2'-deoxyuridine, 2'-amino-2'-deoxy-uridine, uridine, 2'-amino-2'-deoxy, 1-2r,3r,4s,5r-3-amino-4-hydroxy-5-hydroxymethyl oxolan-2-yl-3h-pyrimidine-2,4-dione, 2'-amino,2'-deoxyuridine, 2'-amino-2'-deoxy uridine, 2'-deoxy-2'-amino-uridine, 2'-amino-2'-deoxy-d-uridine, 2,4 1h,3h-pyrimidinedione, 1-2-amino-2-deoxy-b-d-ribofuranosyl
PubChem CID 6482479
IUPAC Name 1-[(2R,3R,4S,5R)-3-amino-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
SMILES C1=CN(C(=O)NC1=O)C2C(C(C(O2)CO)O)N
Specifications
Color White to Yellow
Quantity 1 g
Solubility Information Partly soluble in water.
Formula Weight 243.22
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2'-Amino-2'-deoxyuridine