missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-3-(trifluoromethyl)benzaldehyde, 97%, Thermo Scientific™

Product Code. 15425187
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15425187 5 g 5g
15415187 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15425187

Brand: Thermo Scientific Alfa Aesar H32462.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134741-65-4
Molecular Formula C8H3Cl2F3O
Molecular Weight (g/mol) 243.006
MDL Number MFCD11519344
InChI Key MARVQTJKYZMKTO-UHFFFAOYSA-N
Synonym 2,6-dichloro-3-trifluoromethyl benzaldehyde, 2,6-dichloro-3-trifluoromethyl benzaldehyde, jrd
PubChem CID 14793910
IUPAC Name 2,6-dichloro-3-(trifluoromethyl)benzaldehyde
SMILES C1=CC(=C(C(=C1C(F)(F)F)Cl)C=O)Cl

Specifications

Melting Point 48°C to 50°C
Quantity 5 g
Sensitivity Air Sensitive
Formula Weight 243.01
Percent Purity 97%
Chemical Name or Material 2,6-Dichloro-3-(trifluoromethyl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.