accessibility menu, dialog, popup

UserName

CAS RN 134741-65-4

FFFClOCl

CAS RN 134741-65-4

IUPAC Name: 2,6-dichloro-3-(trifluoromethyl)benzaldehyde
Synonyms: 2,6-dichloro-3-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol): 243.01
Molecular Formula: C8H3Cl2F3O
InChi Key: MARVQTJKYZMKTO-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=C(Cl)C(C=O)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results