missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethoxythiophenol, 95%, Thermo Scientific™

Catalog Number 11302547
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1483-27-8
C8H10O2S
170.23
MFCD00052738
SESUUAOAUZDHHP-UHFFFAOYSA-N
2,5-dimethoxythiophenol, 2,5-dimethoxybenzene-1-thiol, benzenethiol, 2,5-dimethoxy, pubchem6852, acmc-1cgdz, 3,6-dimethoxybenzenethiol, #, 2,5-dimethoxyphenyl mercaptan
519009
COC1=CC(S)=C(OC)C=C1
This item is not returnable. View return policy
1483-27-8
C8H10O2S
170.23
MFCD00052738
SESUUAOAUZDHHP-UHFFFAOYSA-N
2,5-dimethoxythiophenol, 2,5-dimethoxybenzene-1-thiol, benzenethiol, 2,5-dimethoxy, pubchem6852, acmc-1cgdz, 3,6-dimethoxybenzenethiol, #, 2,5-dimethoxyphenyl mercaptan
519009
COC1=CC(S)=C(OC)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 1.14
Boiling Point 278°C
Flash Point 122°C (252°F)
Odor Stench
Refractive Index 1.5845
Quantity 5 g
UN Number UN3334
Beilstein 1867842
Sensitivity Air Sensitive
IUPAC Name 2,5-dimethoxybenzene-1-thiol
Formula Weight 170.23
Percent Purity 95%
Chemical Name or Material 2,5-Dimethoxythiophenol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'