Learn More
2,3,5,6-Tetrafluorobenzyl alcohol, 98%
Product Code. 11436257
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
11436257 25 g 25g
11416257 1 g 1g
11426257 5 g 5g
3 options
Product Code. 11436257
Brand: Thermo Scientific Alfa Aesar B25162.14
Request Bulk or Custom Format
CAS: 4084-38-2 | C7H4F4O | 180.10 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4084-38-2
Molecular Formula C7H4F4O
Molecular Weight (g/mol) 180.10
MDL Number MFCD00792428
InChI Key AGWVQASYTKCTCC-UHFFFAOYSA-N
PubChem CID 2734029
IUPAC Name (2,3,5,6-tetrafluorophenyl)methanol
SMILES OCC1=C(F)C(F)=CC(F)=C1F
Specifications
Melting Point 34°C to 38°C
Boiling Point 220°C to 225°C
Odor Recognizable
Quantity 25 g
Synonym 2,3,5,6-tetrafluorobenzyl alcohol, 2,3,5,6-tetrafluorophenyl methanol, 2,3,5,6-tetrafluorobenzylalcohol, benzenemethanol, 2,3,5,6-tetrafluoro, 2,3,5,6-tetrafluorophenyl methan-1-ol, 2,3,5,6-tetrafluorobenyl alcohol, pubchem15529, acmc-209jh7, 2,3,5,6-tetrafluorbenzyl alcohol, benzenemethanol,2,3,5,6-tetrafluoro
Formula Weight 180.1
Percent Purity 98%
Chemical Name or Material 2,3,5,6-Tetrafluorobenzyl alcohol