CAS RN 4084-38-2
CAS RN 4084-38-2
IUPAC Name:
(2,3,5,6-tetrafluorophenyl)methanol
Synonyms:
(2,3,5,6-tetrafluorophenyl)methanol
Molecular Weight (g/mol):
180.1
Molecular Formula:
C7H4F4O
InChi Key:
AGWVQASYTKCTCC-UHFFFAOYSA-N
SMILES:
OCC1=C(F)C(F)=CC(F)=C1F
1
–
1
of
1
results
1
2,3,5,6-Tetrafluorobenzyl alcohol, 98%
CAS: 4084-38-2 Molecular Formula: C7H4F4O Molecular Weight (g/mol): 180.10 MDL Number: MFCD00792428 InChI Key: AGWVQASYTKCTCC-UHFFFAOYSA-N PubChem CID: 2734029 IUPAC Name: (2,3,5,6-tetrafluorophenyl)methanol SMILES: OCC1=C(F)C(F)=CC(F)=C1F