missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethylhexanoic acid, 94%, Thermo Scientific Chemicals

Product Code. 11313646
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
11313646 250 mg 250mg
11303646 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11313646

Brand: Thermo Scientific Alfa Aesar L14664.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 813-72-9
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.21
MDL Number MFCD00965097
InChI Key YTTWDTVYXAEAJA-UHFFFAOYSA-N
Synonym 2,2-dimethyl-hexanoic acid, hexanoic acid,2,2-dimethyl, 2,2-dimethylhexanoicacid, dimethylhexanoic acid, dimethyl-hexanoic acid, acmc-209plb, 2,2-dimethyl hexanoic acid, #, hexanoic acid, 2,2-dimethyl, yttwdtvyxaeaja-uhfffaoysa
PubChem CID 163217
IUPAC Name 2,2-dimethylhexanoic acid
SMILES CCCCC(C)(C)C(O)=O

Specifications

Boiling Point 216°C to 220°C
Quantity 250 mg
UN Number UN3265
Beilstein 1749961
Formula Weight 144.22
Percent Purity 94%
Chemical Name or Material 2,2-Dimethylhexanoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.