CAS RN 813-72-9
CAS RN 813-72-9
IUPAC Name:
2,2-dimethylhexanoic acid
Synonyms:
2,2-dimethylhexanoic acid
Molecular Weight (g/mol):
144.21
Molecular Formula:
C8H16O2
InChi Key:
YTTWDTVYXAEAJA-UHFFFAOYSA-N
SMILES:
CCCCC(C)(C)C(O)=O
1
–
1
of
1
results
1
2,2-Dimethylhexanoic acid, 94%, Thermo Scientific Chemicals
CAS: 813-72-9 Molecular Formula: C8H16O2 Molecular Weight (g/mol): 144.21 MDL Number: MFCD00965097 InChI Key: YTTWDTVYXAEAJA-UHFFFAOYSA-N Synonym: 2,2-dimethyl-hexanoic acid,hexanoic acid,2,2-dimethyl,2,2-dimethylhexanoicacid,dimethylhexanoic acid,dimethyl-hexanoic acid,acmc-209plb,2,2-dimethyl hexanoic acid,#,hexanoic acid, 2,2-dimethyl,yttwdtvyxaeaja-uhfffaoysa PubChem CID: 163217 IUPAC Name: 2,2-dimethylhexanoic acid SMILES: CCCCC(C)(C)C(O)=O