Learn More
RETSCH 180° Stainless Steel Sieve For SR 200 Rotor Beater Mills
Product Code. 10289061 Shop All Retsch Products
Click to view available options
Pore Size:
0.08 mm
0.12 mm
0.2 mm
0.25 mm
0.5 mm
0.75 mm
1 mm
1.25 mm
1.5 mm
2 mm
3 mm
4 mm
10 mm
Unit Size:
Each
Product Code. 10289061
Brand: RETSCH 03.647.0048
Only null left
Specifications
Quantity 1Each
Pore Size 3 mm
Type Sieve
Material Stainless Steel
For Use With (Equipment) SR 200 Rotor Beater Mills