Learn More
RETSCH 180° Stainless Steel Sieve For SR 200 Rotor Beater Mills
Product Code. 10219211 Shop All Retsch Products
Change view
Click to view available options
Pore Size:
0.08 mm
0.12 mm
0.2 mm
0.25 mm
0.5 mm
0.75 mm
1 mm
1.25 mm
1.5 mm
2 mm
3 mm
4 mm
10 mm
Unit Size:
Each
Product Code. Pore Size unitSize
10219211 10 mm Each
10222691 0.5 mm Each
10126691 0.12 mm Each
10796861 0.2 mm Each
10044761 0.25 mm Each
10700442 0.08 mm Each
10228971 0.75 mm Each
10064771 1.25 mm Each
10238971 1.5 mm Each
10100741 1 mm Each
10165971 2 mm Each
10289061 3 mm Each
10084781 4 mm Each
13 options
Product Code. 10219211
Brand: RETSCH 03.647.0053
Only null left
Specifications
Quantity 1Each
Pore Size 10 mm
Type Sieve
Material Stainless Steel
For Use With (Equipment) SR 200 Rotor Beater Mills