Learn More
RETSCH 180° Stainless Steel Sieve For SR 200 Rotor Beater Mills
Shop All Retsch Products
Click to view available options
Pore Size:
0.08 mm
0.12 mm
0.2 mm
0.25 mm
0.5 mm
0.75 mm
1 mm
1.25 mm
1.5 mm
2 mm
3 mm
4 mm
10 mm
Unit Size:
Each
Specifications
Specifications
| Quantity | 1Each |
| Pore Size | 4 mm |
| Type | Sieve |
| Material | Stainless Steel |
| For Use With (Equipment) | SR 200 Rotor Beater Mills |