Learn More
BINDER™ Temperature and Humidity Calibration Certificate (18 Measuring Points)
For Use With: BINDER units series MKF, KMF, KBF, KBF P, KBF LQC, KBWF, MKFT
Brand: BINDER™ DL30-0318
This item is not returnable.
View return policy
Description
- One humidity point of measurement
- Including certificate
- Humidity measurement point in center of chamber
Specifications
BINDER Chambers Series MKF, KMF, KBF, KBF P, KBF LQC, KBWF, MKFT | |
Temperature and Humidity: 18 measurement points | |
Temperature and Humidity Measurement |
Temperature measurement 18 temperature measuring points and 1 humidity measuring point - including certificate, 18 temperature measuring points and 1 humidity measuring point in center of chamber, at 25°C / 60 % RH or (at specified values - for MKF, KMF, KBF, KBF P, KBF LQC, KBWF, MKFT Series) | |
Calibration Certificate |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
BINDER™ Temperature and Humidity Calibration Certificate (18 Measuring Points) > 18 measuring points; For Use With: MKF, KMF, KBF, KBF P, KBF LQC, KBWF, MKFT series