Learn More
Fisherbrand™ 150 Watt Hand Held Homogenizer
Product Code. 15565809 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Plug Type:
EU Plug
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Type Unit Size
15565809 EU Plug Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15565809
  • ONLINE EXCLUSIVE PRICE
    7690.00 NOK
    7080.57 NOK / Pack of 1
    Save 609.43 NOK
    7% Off
View all promotions
Estimated Shipment: 23-12-2026
Add to basket
Add to basket
Process samples reliably, quickly and efficiently. Fisherbrand™ 150 Watt Hand Held Homogenizer is a high performance, light weight tissue homogenizer.
  • Designed for comfortable hand-held use/can be mounted to motor stand for lengthier processing
  • Interchangeable quick connect rotor stator generator probes provides flexibility to process samples from 200μL to 100mL
  • Stainless steel probes (5mm - 10mm diameter) are designed for long life and are easy to disassemble for cleaning
  • Plastic probes virtually eliminate risk of cross contamination and carryover between samples, can be cleaned for reuse in most of the cases
  • Speed: Variable from 5,000 to 35,000rpm
  • Power: 150w max
  • Sound: <72dB
Certifications
CE
Warranty
One year
TRUSTED_SUSTAINABILITY
Specifications
Speed 5000 to 35,000 rpm
Voltage 220 V
Electrical Requirements 220V EU Cord, 150W
Wattage 125 W
Sample Volume (Metric) 200 μL to 100 mL
Diameter (Metric) 6.35 cm
Height (Metric) 22.8 cm
Certifications/Compliance CE, UKCA
For Use With (Application) Tissue homogenization, PCR analysis, cell disruption, RNA/DNA and protein extractions, and emulsions and suspensions
Includes Tip adapter, tool kit, stand clamp, plastic disposable generator probe samples (2/Pk.)
Packaging Quantity 1 Ea.
Packaging Type Box
Plug Type EU Plug
Weight (Metric) 466 g
Product Type Homogenizer