Learn More
Fisherbrand™ 11 mm Plastic Snap Ring Micro-Vial, Glass Micro Insert, Silanized
Shop All Fisherbrand Products
Click to view available options
Color:
Clear
Unit Size:
Pack of 100
Specifications
Specifications
| Description | TopSert |
| Color | Clear |
| Diameter (Metric) Outer | 11.6 mm |
| Material | Hydrolytic Glass (Class I) |
| Volume (Metric) | 0.35 mL |
| For Use With (Application) | GC, HPLC |
| Neck Style | Snap/Crimp |
| Packaging Type | CleanPack, Polypropylene Box |
| Style | Conical Integrated Insert |
| Volume (Metric) Residual | <4 μL |
| Show More |