missing translation for 'onlineSavingsMsg'
Learn More

N-Nitroso-L-hydroxyproline, TRC

Product Code. 17904125
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17904125 10 mg 10mg
17932705 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17904125 Supplier Toronto Research Chemicals Supplier No. TRCN52860010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30310-80-6
Molecular Formula C5H8N2O4
Molecular Weight (g/mol) 160.13
Synonym (4R)-4-Hydroxy-1-nitroso-L-proline, N-Nitroso-4-hydroxyproline, N-Nitrosohydroxyproline;
SMILES O[C@@H]1C[C@H](N(C1)N=O)C(=O)O

Specifications

InChI Formula InChI=1S/C5H8N2O4/c8-3-1-4(5(9)10)7(2-3)6-11/h3-4,8H,1-2H2,(H,9,10)/t3-,4+/m1/s1
Quantity 10 mg
Formula Weight 160.0484
Shipping Condition Room Temperature
Chemical Name or Material N-Nitroso-L-hydroxyproline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.