CAS RN 30310-80-6
CAS RN 30310-80-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Nitroso-L-hydroxyproline, TRC
CAS: 30310-80-6 Molecular Formula: C5H8N2O4 Molecular Weight (g/mol): 160.13 Synonym: (4R)-4-Hydroxy-1-nitroso-L-proline,N-Nitroso-4-hydroxyproline,N-Nitrosohydroxyproline; SMILES: O[C@@H]1C[C@H](N(C1)N=O)C(=O)O