Learn More
Invitrogen™ 1-pyrenebutanoic acid, succinimidyl ester
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
The amine-reactive 1-pyrenebutanoic acid, succinimidyl ester can be used to create environment-sensitive bioconjugates with this unique fluorophore.
Specifications
Specifications
| Quantity | 100 mg |
| Product Type | 1-Pyrenebutanoic Acid Succinimidyl Ester |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Chemical Reactivity | Amine |
| Shipping Condition | Room Temperature |
| Reactive Moiety | Active Ester, Succinimidyl Ester |
| Label or Dye | Pyrene |
| Label Type | Classic Dyes |