Learn More
1-Methoxy-2-methyl-2-propanol, 98+%
Product Code. 11349196
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Product Code. 11349196
Brand: Thermo Scientific Alfa Aesar L20351.22
Request Bulk or Custom Format
CAS: 3587-64-2 | C5H12O2 | 104.15 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3587-64-2
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
MDL Number MFCD03701584
InChI Key MXUXZWFVAPTPAG-UHFFFAOYSA-N
Synonym 1-methoxy-2-methyl-2-propanol, 2-propanol, 1-methoxy-2-methyl, acmc-20aott, 2-methoxymethyl-2-propanol, 1,1-dimethyl-2-methoxyethanol, 2-propanol,1-methoxy-2-methyl, 1-methoxy-2-methyl-2-propanol #
PubChem CID 77137
IUPAC Name 1-methoxy-2-methylpropan-2-ol
SMILES COCC(C)(C)O
Specifications
Density 0.892
Boiling Point 115°C to 116°C
Flash Point 27°C (80°F)
Refractive Index 1.4044
Quantity 100 g
UN Number UN1987
Formula Weight 104.15
Percent Purity ≥98%
Chemical Name or Material 1-Methoxy-2-methyl-2-propanol