CAS RN 3587-64-2
CAS RN 3587-64-2
IUPAC Name:
1-methoxy-2-methylpropan-2-ol
Synonyms:
1-methoxy-2-methylpropan-2-ol
Molecular Weight (g/mol):
104.15
Molecular Formula:
C5H12O2
InChi Key:
MXUXZWFVAPTPAG-UHFFFAOYSA-N
SMILES:
COCC(C)(C)O
1
–
2
of
2
results
1
1-Methoxy-2-methyl-2-propanol, 98+%
CAS: 3587-64-2 Molecular Formula: C5H12O2 Molecular Weight (g/mol): 104.15 MDL Number: MFCD03701584 InChI Key: MXUXZWFVAPTPAG-UHFFFAOYSA-N Synonym: 1-methoxy-2-methyl-2-propanol,2-propanol, 1-methoxy-2-methyl,acmc-20aott,2-methoxymethyl-2-propanol,1,1-dimethyl-2-methoxyethanol,2-propanol,1-methoxy-2-methyl,1-methoxy-2-methyl-2-propanol # PubChem CID: 77137 IUPAC Name: 1-methoxy-2-methylpropan-2-ol SMILES: COCC(C)(C)O