missing translation for 'onlineSavingsMsg'
Learn More

1,4,8,11-Tetrakis(diethylaminocarbonylmethyl)-1,4,8,11-tetraazacyclotetradecane, Thermo Scientific™

Product Code. 15455115
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15455115 250 mg 250mg
11411290 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15455115

Brand: Thermo Scientific Alfa Aesar H26367.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126320-56-7
Molecular Formula C34H68N8O4
Molecular Weight (g/mol) 652.97
MDL Number MFCD09263304
InChI Key JKQNZLKTODZGSY-UHFFFAOYSA-N
Synonym tetamet2, 1,4,8,11-tetrakis diethylaminocarbonylmethyl-1,4,8,11-tetraazacyclotetradecane, acmc-20ap3f, cyclam-n,n',n',n-tetrakis n,n-diethylacetamide, 1,4,8,11-tetrakis diethylaminocarbonylmethyl-1,4,8,11-tetra, 1,4,8,11-tetraazacyclotetradecane-1,4,8,11-tetrakis n,n-diethylacetamide, 2,2',2,2'-1,4,8,11-tetraazacyclotetradecane-1,4,8,11-tetrayl tetrakis n,n-diethylacetamide, n,n-diethyl-2-4,8,11-tris diethylcarbamoyl methyl-1,4,8,11-tetraazacyclotetradecan-1-yl acetamide
PubChem CID 14434737
IUPAC Name N,N-diethyl-2-[4,8,11-tris[2-(diethylamino)-2-oxoethyl]-1,4,8,11-tetrazacyclotetradec-1-yl]acetamide
SMILES CCN(CC)C(=O)CN1CCCN(CCN(CCCN(CC1)CC(=O)N(CC)CC)CC(=O)N(CC)CC)CC(=O)N(CC)CC

Specifications

Melting Point 144°C to 145°C
Quantity 250 mg
Sensitivity Hygroscopic
Formula Weight 652.97
Chemical Name or Material 1,4,8,11-Tetrakis(diethylaminocarbonylmethyl)-1,4,8,11-tetraazacyclotetradecane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.