accessibility menu, dialog, popup

UserName

CAS RN 126320-56-7

CH3NCH3ONNNNONH3CH3CONCH3CH3ONCH3CH3

CAS RN 126320-56-7

IUPAC Name: N,N-diethyl-2-{4,8,11-tris[(diethylcarbamoyl)methyl]-1,4,8,11-tetraazacyclotetradecan-1-yl}acetamide
Synonyms: N,N-diethyl-2-{4,8,11-tris[(diethylcarbamoyl)methyl]-1,4,8,11-tetraazacyclotetradecan-1-yl}acetamide
Molecular Weight (g/mol): 652.97
Molecular Formula: C34H68N8O4
InChi Key: JKQNZLKTODZGSY-UHFFFAOYSA-N
SMILES: CCN(CC)C(=O)CN1CCCN(CC(=O)N(CC)CC)CCN(CC(=O)N(CC)CC)CCCN(CC(=O)N(CC)CC)CC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results