missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Dimethylaminopropyl)piperazine, 99%, Thermo Scientific™

Product Code. 11480240
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11480240 5 g 5g
11470240 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11480240 Supplier Thermo Scientific Alfa Aesar Supplier No. H50690.06
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 877-96-3
Molecular Formula C9H21N3
Molecular Weight (g/mol) 171.288
MDL Number MFCD00082601
InChI Key YJRGRZJKGMBHIB-UHFFFAOYSA-N
Synonym 1-3-dimethylaminopropyl piperazine, n,n-dimethyl-3-piperazin-1-yl propan-1-amine, dimethyl 3-piperazin-1-yl propyl amine, 1-3-dimethylaminopropyl-piperazine, 1-3-dimethylamino propyl piperazine, 1-3-dimethylamino-propyl-piperazine, 1-piperazinepropanamine,n,n-dimethyl, dimethyl-3-piperazin-1-yl-propyl-amine, n,n-dimethyl-3-1-piperazinyl-1-propanamine, n,n-dimethyl-n-3-1-piperazinyl propyl amine
PubChem CID 2736234
IUPAC Name N,N-dimethyl-3-piperazin-1-ylpropan-1-amine
SMILES CN(C)CCCN1CCNCC1

Specifications

Quantity 5 g
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 171.29
Percent Purity 99%
Chemical Name or Material 1-(3-Dimethylaminopropyl)piperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.