accessibility menu, dialog, popup

UserName

H3CNCH3NNH

CAS RN 877-96-3

IUPAC Name: dimethyl[3-(piperazin-1-yl)propyl]amine
Synonyms: dimethyl[3-(piperazin-1-yl)propyl]amine
Molecular Weight (g/mol): 171.29
Molecular Formula: C9H21N3
InChi Key: YJRGRZJKGMBHIB-UHFFFAOYSA-N
SMILES: CN(C)CCCN1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results