missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Chlorophenoxy)-3-butyn-2-ol, 97%, Thermo Scientific™

Product Code. 11431090
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11431090 25 g 25g
11411090 1 g 1g
11421090 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11431090 Supplier Thermo Scientific Alfa Aesar Supplier No. B25598.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86912-84-7
Molecular Formula C10H9ClO2
Molecular Weight (g/mol) 196.63
MDL Number MFCD09909563
InChI Key CEZMXOXWZYDVOU-UHFFFAOYSA-N
Synonym 1-3-chlorophenoxy but-3-yn-2-ol, 1-3-chlorophenoxy-3-butyn-2-ol
PubChem CID 13145644
IUPAC Name 1-(3-chlorophenoxy)but-3-yn-2-ol
SMILES C#CC(COC1=CC(=CC=C1)Cl)O

Specifications

Melting Point 36°C to 40°C
Quantity 25 g
Formula Weight 196.63
Percent Purity 97%
Chemical Name or Material 1-(3-Chlorophenoxy)-3-butyn-2-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.