accessibility menu, dialog, popup

UserName

HCOClOH

CAS RN 86912-84-7

IUPAC Name: 1-(3-chlorophenoxy)but-3-yn-2-ol
Synonyms: 1-(3-chlorophenoxy)but-3-yn-2-ol
Molecular Weight (g/mol): 196.63
Molecular Formula: C10H9ClO2
InChi Key: CEZMXOXWZYDVOU-UHFFFAOYNA-N
SMILES: OC(COC1=CC(Cl)=CC=C1)C#C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results