missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Benzenetricarboxylic acid hydrate, 99%, Thermo Scientific™

Product Code. 11498890
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11498890 10 g 10g
11478097 5 g 5g
11488097 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11498890 Supplier Thermo Scientific Alfa Aesar Supplier No. A17491.09
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used in the synthesis and characterization and thermal behavior of hemimellitic acid complexes with lanthanides(III).

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 732304-21-1
Molecular Formula C9H6O6
Molecular Weight (g/mol) 210.14
MDL Number MFCD00002468
InChI Key UJMDYLWCYJJYMO-UHFFFAOYSA-N
Synonym 1,2,3-benzenetricarboxylic acid hydrate, hemimellitic acid hydrate, benzene-1,2,3-tricarboxylic acid, oxamethane, acmc-1ajgh
PubChem CID 16219028
SMILES OC(=O)C1=CC=CC(C(O)=O)=C1C(O)=O

Specifications

Melting Point ∼191°C (decomposition)
Quantity 10 g
Beilstein 2214816
Solubility Information Slightly soluble in water.
IUPAC Name benzene-1,2,3-tricarboxylic acid;hydrate
Formula Weight 210.14 (Anhydrous)
Percent Purity 99%
Chemical Name or Material 1,2,3-Benzenetricarboxylic acid hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.