accessibility menu, dialog, popup

UserName

OOHOOHOOH

CAS RN 732304-21-1

IUPAC Name: benzene-1,2,3-tricarboxylic acid hydrate
Synonyms: hemimellitic acid hydrate
Molecular Weight (g/mol): 228.16
Molecular Formula: C9H8O7
InChi Key: DYDNZUYNRZENMU-UHFFFAOYSA-N
SMILES: O.OC(=O)C1=CC=CC(C(O)=O)=C1C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results