Learn More
Thermo Scientific Chemicals 1,2,3,5-Tetra-O-acetyl-beta-L-ribofuranose, 98%
Product Code. 15407299
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15407299 1 g 1g
15417299 5 g 5g
15427299 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15407299 Supplier Thermo Scientific Chemicals Supplier No. J66074.03
 Restricted Item
In Stock
Add to basket
CAS: 144490-03-9 | C13H18O9 | 318.278 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 144490-03-9
Molecular Formula C13H18O9
Molecular Weight (g/mol) 318.278
MDL Number MFCD00674547
InChI Key IHNHAHWGVLXCCI-UMSGYPCISA-N
Synonym 1,2,3,5-tetra-o-acetyl-beta-l-ribofuranose, 1,2,3,5-tetra-o-acetyl-b-l-ribofuranose, 2s,3s,4s,5r-3,4,5-tris acetyloxy oxolan-2-yl methyl acetate, beta-l-ribofuranose 1,2,3,5-tetra-o-acetate, beta-l-ribofuranose tetraacetate, 1,2,3,5-tetra-o-acetyl-?-l-ribofuranose, 2s,3s,4s,5r-3,4,5-triacetyloxyoxolan-2-yl methyl acetate, 2r,3s,4s,5s-5-acetoxymethyl tetrahydrofuran-2,3,4-triyl triacetate
PubChem CID 69431757
IUPAC Name [(2S,3R,4R,5S)-3,4,5-triacetyloxyoxolan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C
Specifications
Melting Point 80°C to 83°C
Color White
Quantity 1 g
Optical Rotation +12.7° (c=2.4 in Chloroform)
Formula Weight 318.3
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1,2,3,5-Tetra-O-acetyl-beta-L-ribofuranose