Learn More
RETSCH 1.0353 Stainless Steel Bottom Sieve For SM 200 Cutting Mills
Brand:  RETSCH 03.647.0359
24647.75 NOK valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 11994322
  • 25945.00 NOK / Each
Estimated Shipment: 21-08-2024
Specifications
1Each
Bottom Sieves
SM 200 Cutting Mill
10mm