Learn More
Eppendorf™ 0.5 Model ThermoMixer™
Product Code. 15316551 Shop All Eppendorf Products
Change view
Click to view available options
Plug Type:
EU
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Type Unit Size
15316551 EU Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15316551 Supplier Eppendorf™ Supplier No. 5386000010
  • ONLINE EXCLUSIVE PRICE
    38100.00 NOK
    35640.06 NOK / Pack of 1
    Save 2459.94 NOK
    6% Off
View all promotions
 Restricted Item
In Stock
Specifications
Accuracy Max. ±0.5°C at 20°C to 45°C
Content And Storage Thermoblock 24 x 0.5 mL, EU
Holds 0.5 mL Microtubes
No. of Places 24
Plug Type EU
Temperature Range (Metric) 1°C to 100°C
For Use With (Application) Used to Transformation of Bacteria Strains and Plasmids
Includes USB Interface
Model ThermoMixer F0.5
Speed 300 to 2500 rpm
Width (Metric) 206 mm
Height (Metric) 163 mm
Depth (Metric) 304 mm
Electrical Requirements 220 to 240 V, 50/60 Hz, 200 W
Voltage 220 to 240 V
Frequency 50 to 60 Hz