Learn More
Noxa Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
Brand:  Novus Biologicals NB100-56368
 View more versions of this product
Product Code. 18796523
  • ONLINE EXCLUSIVE PRICE
    4490.00 NOK
    4246.20 NOK / 0.10mg
    Save 243.80 NOK
    5% Off
Description
Noxa Polyclonal specifically detects Noxa in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Immunofluorescence, Immunohistochemistry (Paraffin)
Q9JM54
Protein G purified
Purified
Polyclonal
adult T cell leukemia-derived PMA-responsive, APR, Immediate-early-response protein APR, NOXAPMA-induced protein 1, phorbol-12-myristate-13-acetate-induced protein 1, PMA-Induced Protein 1, Protein Noxa
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.