Learn More
Noxa Antibody, Novus Biologicals™
Product Code. p-5086766 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
Unit Size:
0.1mg
25µg
Product Code. Quantity unitSize
18706953 0.025 mg 25µg
18796523 - 0.1mg
2 options
Product Code. 18706953
Brand: Novus Biologicals NB10056368SS
Rabbit Polyclonal Antibody has been used in 5 publications
Noxa Polyclonal specifically detects Noxa in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Antigen Noxa
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 2 ug/mL, Immunohistochemistry 1:50 - 1:250, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:50 - 1:250. Use reported in scientific literature (Inta et al. (2006))
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. Q9JM54
Gene Alias adult T cell leukemia-derived PMA-responsive, APR, Immediate-early-response protein APR, NOXAPMA-induced protein 1, phorbol-12-myristate-13-acetate-induced protein 1, PMA-Induced Protein 1, Protein Noxa
Gene Symbols PMAIP1
Host Species Rabbit
Immunogen This Noxa antibody was developed by immunizing rabbits with a mixture of synthetic peptides containing amino acids 51-66 and 75-90 of mouse Noxa.
Molecular Weight of Antigen 11.6 kDa
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5366
Test Specificity Amino acids 51-66 of mouse Noxa are 100% homologous in mouse and 77% in rat; amino acids 75-90 of mouse Noxa are 100% homologous in mouse, 93% homologous in rat and 76% homologous in human.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
For Research Use Only