CAS RN 99675-03-3
CAS RN 99675-03-3
IUPAC Name:
propan-2-yl 2-({methoxy[(propan-2-yl)amino]sulfanylidene-λ⁵-phosphanyl}oxy)benzoate
Synonyms:
isopropyl 2-{[(isopropylamino)(methoxy)sulfanylidene-λ⁵-phosphanyl]oxy}benzoate
Molecular Weight (g/mol):
331.37
Molecular Formula:
C14H22NO4PS
InChi Key:
IXTOWLKEARFCCP-UHFFFAOYNA-N
SMILES:
COP(=S)(NC(C)C)OC1=CC=CC=C1C(=O)OC(C)C
1
–
7
of
7
results
1
Isofenphos-methyl 100 μg/mL in Acetone, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
Methyl Isofenphos (Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Methyl Isofenphos (Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Methyl Isofenphos (Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.